Rods for Thermal Conductivity
# de catalogue 470230-168
Fournisseur: Wards
Spécifications
- Description:Rods for Thermal Conductivity
- Size:11.88L x 0.13" dia. Rod
- Cat. no.:470230-168
Spécifications
A propos de cet article
Various rods to explore thermal conductivity.
- Contains iron, copper, aluminum, lead, brass, and zinc rods
- Used for thermal conductivity experiments
The Thermal Conductivity Rods provide an excellent tool for use in thermal conductivity experiments. Rods made with 6 different metals are included (Iron, Copper, Aluminum, Lead, Brass, and Zinc), making studying thermal conductivity of various metals
Customers Who Bought This Also Bought