Ice Melting Plates
# de catalogue 470230-398
Fournisseur: Wards
Spécifications
- Description:Ice Melting Plates
Spécifications
A propos de cet article
Ideal for studying heat and thermodynamics.
- Kit includes 2 plates: aluminum, plastic foam, each plate is 9 x 9 x 1 cm
- The aluminum plate that is cold to the touch melts the ice faster than the plastic foam plate, even though both have equivalent temperatures
- A great demonstration for conductivity and heat transfer
- Instruction manual provided
This elegantly simple kit challenges student's preconceived notions on concepts of heat and thermodynamics. The melting plates display quizzical properties when an ice cube melts on each plate. The aluminum plate that is cold to the touch melts the ice faster than the plastic foam plate, even though both have equivalent temperatures. A great demonstration for conductivity and heat transfer.
Customers Who Bought This Also Bought