ContactUsLinkComponent
Hermle Universal Benchtop Centrifuges
Hermle Universal Benchtop Centrifuges
# de catalogue 470230-648
Fournisseur:  Ward's
Hermle Universal Benchtop Centrifuges
# de catalogue 470230-648
Fournisseur:  Ward's
Spécifications
  • Description:
    Rotor for 100 ml Tubes
  • Cat. no.:
    470230-648
  • Maximum Speed:
    4500 rpm
  • Maximum RCF:
    3350×g
  • Capacity:
    4 Tubes
Spécifications
A propos de cet article
Setting new standards in this centrifuge class.
  • Bundles are available with our most popular rotor choices
  • Models available with or without refrigeration
  • 19 different rotor choices available to choose from
  • Suitable for nearly every laboratory application
With 19 available rotor options, the Z326 defines the term “universal”. Swinging bucket rotors and high speed angled rotors are available to accept tubes from 0.2 ml up to 85 ml, making this centrifuge suitable for nearly any laboratory application. Despite an impressive capacity of up to 4×100 ml (or 6×85 ml), the unit is surprisingly compact with both the standard air-cooled model and refrigerated model featuring a width of just over 15".
Advanced microprocessor controls allow for instant rotor identification, which automatically converts the rotational speed to the relevant g-force (rcf), preventing the user from setting a speed above the capabilities of the installed rotor. In addition to retrieving saved programs, the easy to use front panel can adjust accel/decel rates, and change the end of run alarm tone.
Customers Who Bought This Also Bought