ContactUsLinkComponent
Digital to Analogue Convertor
Digital to Analogue Convertor
# de catalogue 470230-450
Fournisseur:  Wards
Digital To Analogue Convertor
Digital to Analogue Convertor
# de catalogue 470230-450
Fournisseur:  Wards
Spécifications
  • Description:
    Digital to Analogue Convertor
  • Cat. no.:
    470230-450
  • Dimensions:
    8×5×2.25"
Spécifications
A propos de cet article
Demonstrates basis of digital communications.
  • Converts digital signal to an analog output
  • Great for demonstrating the basis of digital communications
  • Companion apparatus to the labs analog to digital converter
This is great companion unit to the Labs Analog to Digital Converter (PH1366). A 3 bit binary input (0 V represents 0 and 5 V represents 1) is converted to 0 to 7 V analogue output. By using the two units together the sampling process can be demonstrated to show that information is lost during the conversion process. The Digital to Analog Converter can be powered from the ADC or from a separate 12 V power supply.
Customers Who Bought This Also Bought