Energy Conversion Kit
# de catalogue 470230-446
Fournisseur: Wards
Spécifications
- Description:Energy Conversion Kit
- Cat. no.:470230-446
- Dimensions:10×8×6.75" (apparatus)
Spécifications
A propos de cet article
Demonstrates process of energy conversion.
- Great tool to discover conversion processes of different types of energy
- Includes 3 different devices to generate electricity
- Includes 5 devices that converts the generated electricity into 5 different forms of energy
- Perfect for hands on experience in the classroom
This kit wonderfully demonstrates the process of energy conversion between many different forms of energy. Ideal for classroom demonstrations as well as a hands on learning tool. Mounted to a solid plastic board are three different sources for the generation of energy: hand crank (mechanical), cell battery holder (electrical), and a solar cell (light). Five circuit elements can each be inserted into the board to observe the use of the generated electrical energy: motor with fan (mechanical), buzzer (sound), LED (light), resistor (heat), and an electromagnet (magnetism). Uses 2 'C' batteries (not included).
Ordering information: Uses 2 'C' batteries (not included).
Customers Who Bought This Also Bought