St. Louis Motor Basic
# de catalogue 470230-434
Fournisseur: Wards
Spécifications
- Description:St. Louis Motor Basic
- Cat. no.:470230-434
Spécifications
A propos de cet article
Demonstrates operation of motors and generators.
- St Louis motor demonstration is useful in demonstrating the operations of motor and generator
- Consisting of a two-pole armature mounted between magnets
- Steel supports are mounted on a ridged 4" dia. plastic base
- Will operate on 3 to 6 V DC
- Power supply not included
St Louis Motor Demonstration is useful in demonstrating the operations of motor and generator. Consisting of a two-pole armature mounted between magnets. Steel supports are mounted on a ridged 4" Diameter plastic base. Will operate on 3 to 6 V DC. Power supply not included. This open design allows students to see each part of the motor operate, which shows basic function.
Customers Who Bought This Also Bought