VWR® Weighing Bottles with Stopper, Low Form, Cylindrical
Supplier: VWR International
Certificates
About this item
These low form weighing bottles are used for precise weighing of solids.
- Complies ASTM - 438, TYPE-1, Boro 3.3 glass
- These bottles are chemically resistant and mechanically strong
These weighing bottles come with a ground-plug style glass stopper which seals the contents to prevent spilling.
Manufactured from 3.3 expansion ASTM 438 Type 1 Class A standard borosilicate glass.
Customers Who Bought This Also Bought