Capacitance Substitution Box
# de catalogue 470230-408
Fournisseur: Wards
Spécifications
- Description:Capacitance Substitution Box
- Cat. no.:470230-408
Spécifications
A propos de cet article
Ideal for general purpose electronic work.
- Uses 1 rotary dial with 12 steps
- Individual values are 100pF, 220 pF, 470 pF, 2.2 nF, 4.7 nF, 10 nF, 22 nF, 47 nF, 100nF, 220 nF, 470 nF, 1 mFd (1 µF)
- All values are + 5% accuracy and of 63V ratings
Capacitor Box, using 1 rotary dial with 12 steps. Various capacitors are selected one by one. Useful for general purpose electronic work. The unit has an easily selectable range of 12 preferred capacitors from 100 pF to 1 mFd. The individual values are 100 pF, 220 pF, 470 pF, 2.2 nF, 4.7 nF, 10 nF, 22 nF, 47 nF, 100 nF, 220 nF, 470 nF and 1 mFd (1 µF). All values of + 5% Accuracy and of 63V ratings.
Customers Who Bought This Also Bought