Buzzer
# de catalogue 470230-386
Fournisseur: Wards
Spécifications
- Description:Buzzer
- Cat. no.:470230-386
- Dimensions:2.875×2.875" (board)
Spécifications
A propos de cet article
Simple circuit module.
- Simple Circuit module can be used with Worcester Circuit board
- Has two threaded 4 mm terminals with nut to securely hole connectors.
- Can be used with Worcester Circuit Board Kit ( PH1301)
- Low Current buzzer approx. 2mA
Simple Circuit module can be used with Worcester Circuit board, Low Current buzzer approx. 2mA. Can be used with Worcester Circuit Board Kit ( PH1301), Has two 4 mm terminals. 2.875" x 2.875" Board. Equipped with zip tie holders on the back of the board.
Customers Who Bought This Also Bought