Miniature Lamp Bulbs
# de catalogue 470230-376
Fournisseur: Wards
Spécifications
- Description:Lamp bulbs, round 1.5 V, pack 10
- Cat. no.:470230-376
Spécifications
A propos de cet article
Replacement flash lamp bulbs.
- Size E10 Screw Base
- For use with basic circuit kits
- Works with flash lamps, spot lamps and panel lamps
- Pack of 10
Flash Lamp Bulb - 1.5 Volts with M.E.S. cap - For Flash Lamps, Spot Lamps and Panel Lamps and for use in circuit kits. Pack of 10.
Customers Who Bought This Also Bought