High-Intensity Egg Candler | Avantor
High-Intensity Egg Candler
Egg Candler
Catalog # 470019-372
Supplier:   Brinsea Products
Fournisseur #:  USF190
High-Intensity Egg Candler
Catalog # 470019-372
Supplier:  Brinsea Products
High-Intensity Egg Candler
High-Intensity Egg Candler
Egg Candler
Price
$101.10
Log in pour la disponibilité
Each
Product Details & Documents
Handheld and easy to use.
  • High intensity LED light
  • Two sizes of rings accommodate the most common eggs
The OvaView High Intensity egg candler has a higher light output from a higher spec LED than the standard OvaView candling light.
It is designed to be either hand-held or can be left on a work surface. Battery powered for convenience the OvaView High Intensity candling lamp uses high output high efficiency LED illumination – so no bulbs to replace and no concerns about overheating the eggs (even very small eggs) during inspection. Supplied with both the normal flexible black ring plus a special smaller ring for smaller eggs.
Ordering information: Requires 4 AA batteries (not included). Due to the high intensity bulb, rechargeable batteries are recommended.
Spécifications
  • Désignation:
    Egg Candler