Cell Holders with Terminals on Base
# de catalogue 470230-350
Fournisseur: Wards
Spécifications
- Description:Cell Holders with Terminals on Base
- Cat. no.:470230-350
Spécifications
A propos de cet article
Cell holder for simple circuit.
- Base is made of folded sheet metal, sockets are RoHS compliant high quality 4 mm banana plug compatible
- Perfect for constructing simple circuits
The perfect piece of equipment to create power for simple circuits using 4mm banana plugs. Spring loaded connection terminals and secure battery snaps hold (2) D batteries to provide power for any circuit that you build.
Customers Who Bought This Also Bought