ContactUsLinkComponent
Needle Electroscope
Needle Electroscope
# de catalogue 470230-318
Fournisseur:  Wards
Electroscope Needle
Needle Electroscope
# de catalogue 470230-318
Fournisseur:  Wards
Spécifications
  • Description:
    Needle Electroscope
  • Cat. no.:
    470230-318
Spécifications
A propos de cet article
Ideal for experiments with electrostatic principles.
  • Great tool to discover electrostatic principles such as conduction, induction and coulomb repulsion
  • Clear front pane lifts up to allow easy application of ionizing materials
  • Perfect for hands on experience in the classroom
This electroscope is great for hands on experience with electrostatic principles in physics classrooms and labs. The sheet metal walls of the electroscope are attached to a plastic base and top. Front glass window slides up to allow insertion of ionizing material in chamber. A light-weight metal vane is used as a pointer in the electroscope. This light weight metal blade deflects when the electroscope plate is charged. A graduated scale is provided to measure the angle of the deflection. Fitted with 4mm socket. Assembled, the electroscope measures 4 x 3.75 x 2.25 inches.
Customers Who Bought This Also Bought