ContactUsLinkComponent
Electroscope Flask with Aluminum Leaf
Electroscope Flask with Aluminum Leaf
# de catalogue 470230-316
Fournisseur:  Wards
Electroscope - Aluminum Leaf
Electroscope Flask with Aluminum Leaf
# de catalogue 470230-316
Fournisseur:  Wards
Spécifications
  • Description:
    Electroscope Flask with Aluminum Leaf
  • Cat. no.:
    470230-316
Spécifications
A propos de cet article
Demonstrates observation of polarity.
  • 7.5" height with 3.5" dia.
  • Glass conical flask
  • Brass rod and aluminum leaf
  • Detect electric charges and determine polarity
Aluminum leaf electroscope used to observe polarity through electric charge detection. It is comprised of a glass conical flask with a brass rod, disc and aluminum leaf supported by a rubber cork.
Customers Who Bought This Also Bought