Electroscope Flask with Aluminum Leaf
# de catalogue 470230-316
Fournisseur: Wards
Spécifications
- Description:Electroscope Flask with Aluminum Leaf
- Cat. no.:470230-316
Spécifications
A propos de cet article
Demonstrates observation of polarity.
- 7.5" height with 3.5" dia.
- Glass conical flask
- Brass rod and aluminum leaf
- Detect electric charges and determine polarity
Aluminum leaf electroscope used to observe polarity through electric charge detection. It is comprised of a glass conical flask with a brass rod, disc and aluminum leaf supported by a rubber cork.
Customers Who Bought This Also Bought