LED Array
# de catalogue 470230-066
Fournisseur: Wards
Spécifications
- Description:LED array
- Cat. no.:470230-066
Spécifications
A propos de cet article
The unit features 11 vertically stacked colors ranging from bright blue to white.
- Use to study color vs wavelength and/or experimentally determine Planck's Constant h
- Works well with Labs Direct Vision Spectroscope (product number PH100QA)
- 11 vertically stacked LED lights
- Plot wavelength vs threshold voltage to experimentally determine Planck's Constant
- 4 mm banana plugs/power supply NOT included. Operates using 0-6V continuous DC unit (Labs part # EPR1332)
LED Array for use in topics of diffraction gratings and light, as well as Planck's Constant calculation. The unit features 11 vertically stacked colors ranging from bright blue to white and is a versatile classroom tool.
Customers Who Bought This Also Bought