Bourdon Gauge
# de catalogue 470230-056
Fournisseur: Wards
Spécifications
- Description:Bourdon Gauge
- Height:8.31"
- Cat. no.:470230-056
- I.D.:3.88" Gauge Face;
4" Base
Spécifications
A propos de cet article
A working gauge is mounted in a transparent case so the actual working parts are visible.
- Black base with gauge and nipple (0.340in dia.)
- Gauge enclosed in clear plastic case to allow inner workings to be visible
- Kg/cm2 delineated with black markings and psi delineated with red markings
The Bourdon gauge is a safe and simple tool for measuring the pressure of various gases and liquids. The stand has a nipple with which to connect a gas or liquid source hose and its easy to read gauge makes it very user-friendly.
Customers Who Bought This Also Bought