Density, Set Assorted Shapes, Set 14
# de catalogue 470230-054
Fournisseur: Wards
Spécifications
- Description:Density Set, Assorted Shapes, Set 14
- Cat. no.:470230-054
Spécifications
A propos de cet article
Fourteen materials in the form of rectangular blocks.
- 14 different blocks
- Kit contains 11 different materials,
- This kit is intended to familiarize students with appearance, feel, texture and density of these common materials.
- All measurements are approx. and may vary from kit to kit
Ordering information: Kit consists of 14 blocks: Aluminum Block (2" x 1.5" x 1.125"); Aluminum Rectangle (3.875" x 0.75" x 0.75"); Brass Block (2" x 0.75" x 0.75"); Styrofoam block (2" x 1.5" x 1.125"); Wax Block (2" x 1.5" 1.125"); Stainless Steel Block (2" x 2" x 0.75”); Wood Block (2" x 1.5625" x 1.1875”); Wood Rectangle (3.875" x 0.75" x 0.75"); Soap stone Rectangle (3.875" x 0.75" x 0.75"); Marble Rectangle (3.875" x 0.75" x 0.75"); Glass Rectangle (3.875" x 0.75" x 0.75"); Acrylic Rectangle (3.875" x 0.75" x 0.75"); Steel Block (1.5" x 1.5" x 0.75"); Steel Block (2" x 1.5" x 1.125").
Customers Who Bought This Also Bought