Order Entry
ContactUsLinkComponent
Eddy Current Demonstration
Eddy Current Demonstration
# de catalogue 470230-300
Fournisseur:  Wards
Eddy Current Demonstration
Eddy Current Demonstration
# de catalogue 470230-300
Fournisseur:  Wards

Spécifications

  • Description:
    Eddy Current Demonstration
  • Cat. no.:
    470230-300

Spécifications

A propos de cet article

Eddy current for demonstrating Lenz's law.

  • Labs 13" eddy current demonstration
  • Heavy duty 13" copper tube
  • Includes 2 masses, one is steel, one is a neodymium magnet
  • These currents oppose downward motion through a moving magnetic field created by the falling neodymium magnet

Labs 13 eddy current demonstration. This demonstrates the amazing Lenz's law. Consists two masses drop down a 33cm copper tube. One is steel, one is a neodymium magnet. Two masses drop down a 33cm copper tube. One is steel, one is a neodymium magnet. When students see the magnetic mass falls more slowly than the steel mass, you can introduce them to electrical eddy currents. These currents oppose downward motion through a moving magnetic field created by the falling neodymium magnet.

Customers Who Bought This Also Bought