Metal shapes, Set of 12
# de catalogue 470230-046
Fournisseur: Wards
Spécifications
- Description:Metal Shapes, Set of 12
- Cat. no.:470230-046
Spécifications
A propos de cet article
For density determinations, set of twelve in assorted sizes, shapes and masses.
- 12 objects in assorted shapes, sizes and masses
- Brass, iron, aluminum, copper, zinc, lead
- Mass range of 12 - 25 grams
The Labs Set of Metal Blocks is used for density determinations. It includes two of each brass, iron, aluminum, copper, zinc and lead. Masses vary from 12-25 grams.
Customers Who Bought This Also Bought