Equal Mass, Set 5 cylinders lead free
# de catalogue 470230-044
Fournisseur: Wards
Spécifications
- Description:Equal Mass Set, 5 Cylinders lead free
- Cat. no.:470230-044
Spécifications
A propos de cet article
To study the relationship between mass and density. Lead free set.
- 5 Cylinders, 0.5 inch dia..
- Aluminum, Copper, Tin, Zinc, Brass
Equal mass cylinder set contains 5 pieces. 0.50" diameter with different lengths. Aluminum, Copper, Tin, Zinc, Brass.
Customers Who Bought This Also Bought