Order Entry
ContactUsLinkComponent
Solenoid, Air Cored
Solenoid, Air Cored
# de catalogue 470230-276
Fournisseur:  Wards
Solenoid Air Cored
Solenoid, Air Cored
# de catalogue 470230-276
Fournisseur:  Wards

Spécifications

  • Description:
    Solenoid, Air Cored
  • Cat. no.:
    470230-276

Spécifications

A propos de cet article

Solenoid for demonstrating magnetism and electromagnetism.

  • Labs Solenoid Air Cored - Magnetism and Electromagnetism
  • Labs 15" Solenoid is wound with 270 turns of heavy gauge enameled copper wire of 1.25 mm diameter
  • Equipped with 4 mm sockets for power supply.
  • dia. 50 mm 270 turns wire 1.25 mm in dia..
  • Power Supply not included

Labs 15" Solenoid is wound with 270 turns of heavy gauge enameled copper wire of 1.25 mm dia. Equipped with 4 mm sockets for power supply. Diameter 50 mm 270 turns wire 1.25 mm in diameter. Max. Current : 6A Power Supply on 2 safety sockets. The square ends provide stabilization when using for experiments.

Customers Who Bought This Also Bought