Magnetic Field Demonstrator, 4 Plates
# de catalogue 470230-272
Fournisseur: Wards
Spécifications
- Description:Magnetic field Demonstrator, 4 Plates
- Cat. no.:470230-272
Spécifications
A propos de cet article
4 Transparent plates to demonstrate magnetic fields.
- 4 transparent plates to demonstrate magnet fields
- Each plate is 15.3 cm x 7.6 cm x 0.6 cm - 98 chambers
Comprises a set of four transparent plastic plates 153 x 76 x 6 mm, each plate has a matrix of 98 (14 x 7) small circular chambers, each chamber containing an iron rod and functioning effectively as a plotting compass. The plates may be arranged in various ways, e.g. as a single large two dimensional surface, as faces of a cube etc., and in this way may be used to show configuration of a magnetic field in one or two dimensions. The transparent nature of the plates also makes them suitable for use on an overhead projector. Magnet not included. Picture shows only 2 plates - this actually includes 4 plates.
Customers Who Bought This Also Bought