Equal Mass, Set 5 cylinders
# de catalogue 470230-042
Fournisseur: Wards
Spécifications
- Description:Equal Mass Set, 5 Cylinders
- Cat. no.:470230-042
Spécifications
A propos de cet article
To study the relationship between mass and density.
- 5 Cylinders, 0.5 inch dia..
- Aluminum, Copper, Tin, Zinc, Lead
To study the relationship between mass and density. Set of 5 cylinders having same mass and diameter 0.50" varying lengths.
Customers Who Bought This Also Bought