ContactUsLinkComponent
Lenz’ Law
Lenz’ Law
# de catalogue 470230-270
Fournisseur:  Wards
Lenz' Law - Open & Closed Loop
Lenz’ Law
# de catalogue 470230-270
Fournisseur:  Wards
Spécifications
  • Description:
    Lenz’ Law
  • Cat. no.:
    470230-270
Spécifications
A propos de cet article
Loop apparatus ideal for studying Lenz Law and Faraday's Law.
  • Blue cast iron base with rod and needle for holding loops
  • Open loop dimensions: Inner dia. 1.82"/Outer dia. 1.98"
  • Closed loop dimensions: Inner dia. 1.82"/Outer dia. 1.98"
  • Aluminum loops
  • Loops connected by straight bar with groove for placement on needle
The Lenz' Law apparatus with both open and closed loops is effective for studying and demonstrating both Lenz' Law and Faraday's Law.
Customers Who Bought This Also Bought