Metal Cubes, Set for Density
# de catalogue 470230-030
Fournisseur: Wards
Spécifications
- Description:Metal Cubes Set for Density
- Cat. no.:470230-030
Spécifications
A propos de cet article
Density Cubes, Set of 6 Metals, 0.8" (20mm) sides.
- Brass, lead, zinc, copper, aluminum, and iron cubes
- 20mm cubes
The set consists of six 20mm cubes of the following metals: brass, lead, zinc, copper, aluminum, and iron. Perfect for investigating density.
Customers Who Bought This Also Bought