ContactUsLinkComponent
Force Oscillation Resonance
Force Oscillation Resonance
# de catalogue 470229-996
Fournisseur:  Wards
Forced Oscillation And Resonance Demonstrator
Force Oscillation Resonance
# de catalogue 470229-996
Fournisseur:  Wards
Spécifications
  • Description:
    Force Oscillation Resonance
  • Cat. no.:
    470229-996
Spécifications
A propos de cet article
Set it up in under 5 minutes and then demonstrate these difficult concepts with ease.
  • 6V driving motor drives cams attached to spring actuators
  • 6V power supply needed
  • 4 springs, 2 hanging masses, instruction set, and apparatus are included.
  • Demonstrate Hooke's Law, Resonance, Simple Harmonic Motion
Resonance and simple harmonic motion are often difficult to mechanize and then demonstrate, not the case with this device. Set it up in under 5 minutes and then demonstrate these difficult concepts with ease.
Customers Who Bought This Also Bought