VWR® Volumetric Flask, Clear Glass, with Red Vertical Stripe and Glass Penny Head Stopper, Class A
Supplier: VWR International
Certificates
About this item
Flasks are lot certified and supplied with a glass penny head stopper
- Made from Borosilicate 3.3 expansion glass
- Calibrated 'to Contain'
- Batch Certificate included
The red stripe makes it easier to see the meniscus.
Customers Who Bought This Also Bought