Gas Law Demonstration
# de catalogue 470229-968
Fournisseur: Wards
Spécifications
- Description:Gas Law Demonstration
- Cat. no.:470229-968
Spécifications
A propos de cet article
Investigate the dependence of pressure on a gas with changes in temperature and volume.
- Gas Law apparatus with 55ml gas volume
- 50ml syringe
- Rubber stopper
- Three tea candles
- Instructions
Investigate the dependence of pressure on a gas with changes in temperature and volume.
Customers Who Bought This Also Bought