ContactUsLinkComponent
Gas Law Demonstration
Gas Law Demonstration
# de catalogue 470229-968
Fournisseur:  Wards
Gas Law Device For Neulog Sensors
Gas Law Demonstration
# de catalogue 470229-968
Fournisseur:  Wards
Spécifications
  • Description:
    Gas Law Demonstration
  • Cat. no.:
    470229-968
Spécifications
A propos de cet article
Investigate the dependence of pressure on a gas with changes in temperature and volume.
  • Gas Law apparatus with 55ml gas volume
  • 50ml syringe
  • Rubber stopper
  • Three tea candles
  • Instructions
Investigate the dependence of pressure on a gas with changes in temperature and volume.
Customers Who Bought This Also Bought