NZY Broth Dehydrated Media, Teknova
Fournisseur: Teknova
Certificats
A propos de cet article
For use in the propagation of bacteriophage lambda.
- Use 20 grams per liter to reconstitute media
Contains 1.0% Select peptone (NZ amine), 0.5% sodium chloride, 0.5% yeast extract.
Ambient shipping conditions. Store at room temperature.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.