A propos de cet article
For the growth of E. coli and other enteric bacteria.
- Add 35 grams per 1 Liter of water to reconstitute
- Autoclave to sterilize
LB Lennox contains less salt than the most common formulation of LB, known as LB Miller
Ingredients: 1.0% Tryptone (10.0g/L), 0.5% Yeast Extract (5.0g/L), 0.5% NaCl (5.0g/L), 1.5% Agar (15.0g/L).
Ambient shipping conditions. Store at room temperature.
LB Lennox is commonly used for cloning in recombinant E. coli and selection with salt-sensitive antibiotics such as Blasticidin, CloNat, Hygromycin, Puromycin and Zeocin.