A propos de cet article
With full visible and UVA ICH Q1B exposures in as little as 42 hours, the 7450 series provides results fast. Many competing chambers take 2 to 3 times as long. Fast photoexposure allows users to test more samples, reducing drug development time, and saving money.
- Highly reflective specular aluminum interior panels and separate UV and visible light banks create edge-to-edge light uniformity, with over 75% of shelf space usable
- Separate UV and visible Q1B Option II bulbs provide speed without risking exposure overshoot
- Tight temperature uniformity prevents thermal effects from skewing test results.
- Offers a 5 °C operating temperature option, for testing of refrigerated drugs, vaccines, and biologicals
The 7540 Series is built to be facility-friendly, with an easy to use touchscreen controller for "no manual" setting of even the most unique lighting parameters. Slide-out light banks and electrical modules are easy to service, and standard access ports make validation simple. A compact self-contained 7.9 sq.ft benchtop-ready footprint fits well in space-constrained labs.
115 V, 60 Hz model
Light uniformity ±15% VIS (±10% UVA)
UVA light intensity 0 W/M2
VIS light intensity 20 KLux
230 V, 60 Hz model
Light uniformity ±10% VIS (±10% UVA)
UVA light intensity 2 - 30 W/M2
VIS light intensity 2 - 35 Klux
Spécifications
- Catalog No:
- 76102-344
- 76102-342
- 76102-348
- 76102-346
- 76102-352
- 76102-350
- Model:
- 76102-344
- 76102-342
- 76102-348
- 76102-346
- 76102-352
- 76102-350
- Humidity range:
- —
- —
- 40 - 70% RH
- —
- 40 - 70% RH
- 40 - 70% RH
- Int. dimensions:
- 68.3W x 61D x 71.1H cm (26.9 x 24 x 28")
- 68.3W x 61D x 71.1H cm (26.9 x 24 x 28")
- 68.3W x 61D x 71.1H cm (26.9 x 24 x 28")
- 68.3W x 61D x 71.1H cm (26.9 x 24 x 28")
- 68.3W x 61D x 71.1H cm (26.9 x 24 x 28")
- 68.3W x 61D x 71.1H cm (26.9 x 24 x 28")
- Exterior dimensions:
- 95.5W x 76.7D x 113.3H cm (37.6 x 30.2 x 45")
- 95.5W x 76.7D x 113.3H cm (37.6 x 30.2 x 45")
- 95.5W x 76.7D x 113.3H cm (37.6 x 30.2 x 45")
- 95.5W x 76.7D x 113.3H cm (37.6 x 30.2 x 45")
- 95.5W x 76.7D x 113.3H cm (37.6 x 30.2 x 45")
- 95.5W x 76.7D x 113.3H cm (37.6 x 30.2 x 45")
- Time to Results:
- 35 hrs. to ICH 1.2 mil. Lux-hrs./7 hrs. to ICH 200 W-hr./m²
- 60 hrs. to ICH 1.2 mil. Lux-hrs./10 hrs. to ICH 200 W-hr./m²
- 60 hrs. to ICH 1.2 mil. Lux-hrs./10 hrs. to ICH 200 W-hr./m²
- 35 hrs. to ICH 1.2 mil. Lux-hrs./7 hrs. to ICH 200 W-hr./m²
- 35 hrs. to ICH 1.2 mil. Lux-hrs./7 hrs. to ICH 200 W-hr./m²
- 35 hrs. to ICH 1.2 mil. Lux-hrs./7 hrs. to ICH 200 W-hr./m²
- Volume:
- 296 L (10.5 cu.ft.)
- 296 L (10.5 cu.ft.)
- 296 L (10.5 cu.ft.)
- 296 L (10.5 cu.ft.)
- 296 L (10.5 cu.ft.)
- 296 L (10.5 cu.ft.)
- Shipping weight:
- 250 kg (550 lbs.)
- 250 kg (550 lbs.)
- 250 kg (550 lbs.)
- 299 kg (660 lbs.)
- 299 kg (660 lbs.)
- 250 kg (550 lbs.)