VWR® Cell Culture Chamber, Sterile
Fournisseur: VWR International
Certificats
A propos de cet article
This sterile cell culture chamber, which is available with 1, 2, 5, or 10 layers, produces large-scale cell culture results in just one cycle.
- Sterilized by irradiation
- Non-pyrogenic and Dnase-/RNase-free
The device is assembled by hydrophobic membranes with an aperture of 0.22 µm to avoid potential contamination in gas exchange. The device does not absorb cytokines and growth factors, and the conventional trypsin digestion method is applied in cell harvesting. Two 26 mm diameter filling ports allow direct access to the chamber bottom, providing greater flexibility for sterile filling and emptying by pouring, pipetting, or via tubing in a fully closed system. Strict integrity tested.
Certifications: ISO 13485:2003 and ISO 9001:2008.
Customers Who Bought This Also Bought