Outdoor Weather-Resistant Polymer Label, Silver, 3" Core, Brady
# de catalogue 76000-840
Fournisseur: Brady Worldwide
Spécifications
- For Use With:R6600 printer ribbons
- Color:Silver
- Description:Outdoor weather-resistant labels
- Material:Polyester
- Cat. no.:76000-840
- Core Size:3"
- Dimensions:25.4×50.8 mm (1×2")
Spécifications
A propos de cet article
The B-8591 material is an extremely durable, print-on-demand thermal transfer label solution that resists UV, abrasion, and fluids in outdoor use.
- 12+ year outdoor durability reduces identification failures and the costs associated with losing this information
- No over-laminate means durability is drastically improved, without adding an over-laminate over the print
- Quick and easy application with pressure-sensitive adhesive
Certifications: Meets regulatory specifications for halogen-free applications.
Customers Who Bought This Also Bought