Safety Lock and Tag Carrier, Brady Worldwide
Fournisseur: Brady Worldwide
Certificats
A propos de cet article
The Safety Lock and Tag Carrier facilitates quick and easy transportation of lockout and tagout devices to help secure energy isolation points throughout any facility.
- The carrier holds up to 12 safety padlocks and easily attaches to belt clips
- Offset design also permits flush wall or machine mounting
- Includes carabineers and coiled key strap for holding tags, hasps, and keys
Customers Who Bought This Also Bought