Base Excision Repair Antibody Panel 2 Kit, Gene Tex
# de catalogue 89329-734
Fournisseur: Genetex
Numéro CAS:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Description:Base Excision Repair Antibody Panel 2 Kit
- Cat. no.:89329-734
- Supplier No.:GTX300021
Spécifications
A propos de cet article
These sampler kits with combinations of high quality antibodies are convenient and efficient tools to study the mechanism of DNA repair.
Delivery: Kit includes 25 µL each of APE1 antibody [N1] (N-term), PARP antibody [N1N2] (N-term), XRCC1 antibody [N1N3], PNK antibody [N1C1], DNA ligase III antibody [C2C3] (C-term) and Rabbit IgG antibody (HRP).
Caution: Not for any clinical, therapeutic, or diagnostic use in humans or animals. Not for animal or human consumption.
Customers Who Bought This Also Bought