Spécifications
- Description:LB Agar with 100 µg/mL Ampicillin, 60 µg/mL X-Gal, and 0.1 MM IPTG
- Size:100 mm
- Cat. no.:CA100219-318
- Supplier No.:L1902
Spécifications
A propos de cet article
For use with ampicillin resistant strains and strains harboring plasmids such as pBluescript, pGEM, pUC series plasmids. X-gal allows blue/white selection of colonies.
- Sleeve of 20 plates
- Sterile
Contains 1% Tryptone, 0.5% Yeast Extract, 1.0% Sodium Chloride, 1.5% agar 100 µg/ml ampicillin, 0.1 mM IPTG, 60 µg/ml X-gal.
Ambient shipping conditions. Refrigerate to store.