Order Entry
ContactUsLinkComponent
LB Agar Plates with Chloramphenicol 50, Teknova
LB Agar Plates with Chloramphenicol 50, Teknova
Catalog # CA100216-576
Supplier:  Teknova
CAS Number:  
LB Agar Plates with Chloramphenicol 50, Teknova
Catalog # CA100216-576
Supplier:  Teknova
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    LB Agar with 50 µg/mL Chloramphenicol
  • Size:
    100 mm
  • Cat. no.:
    CA100216-576
  • Supplier No.:
    L1019

Specifications

About this item

For use with chloramphenicol resistant strains and strains harboring plasmids such as pBC KS+/-, pJAK17, pMMB208 series plasmids.

  • Sleeve of 20 plates
  • Sterile

Contains 1% Tryptone, 0.5% Yeast Extract, 1.0% Sodium Chloride, 1.5% agar 50 µg/ml chloramphenicol.

Ambient shipping conditions. Refrigerate to store.

Customers Who Bought This Also Bought