SP Wilmad-LabGlass Three-Neck O-Ring Flange Reaction Vessel Covers, SP Industries
Fournisseur: Avantor
Certificats
A propos de cet article
Grooved to accommodate an O-ring and create a greaseless vacuum-tight seal, these borosilicate glass reaction vessel lids have three necks with side necks that are 180° apart.
- All three necks have Standard Taper joints
- Designed to be used with the LG-8071, LG-8079A, LG-8079B, and LG-8079C vessels.
- Flange O.D. is 5.375"
O-ring size: 348
Caution: NOTE: Use clamp LG-7316-106 with these kettle covers.
Customers Who Bought This Also Bought