Ward's® Essentials Crawling Color Walking Water
# de catalogue 470231-282
Fournisseur: Science First
Spécifications
- Description:Crawling Color Walking Water
- Cat. no.:470231-282
Spécifications
A propos de cet article
Explore properties of light with primary and secondary colors.
- Durable acrylic display case for demonstration
- Student-directed inquiry experiments
- Aligned with NGSS MS-PS4, MS-LS1
This high-quality acrylic display allows the classroom to explore color mixing and the properties of water, including cohesion, adhesion, and capillary action. Students take their knowledge a step further by engineering a plant model and mapping capillary action through the “xylem.” This kit includes enough materials for 15 student groups.
Ordering information: Kit includes acrylic setup, food coloring, mixing sheets, disposable pipets, and teacher’s guide.
Customers Who Bought This Also Bought