Muscular Microanatomy Bundle
# de catalogue 470230-944
Fournisseur: Avantor
Spécifications
- Description:Muscle Bundle
- Cat. no.:470230-944
Spécifications
A propos de cet article
Complete Your Muscular Lessons!
- Buy together and save
- 3-year manufacturers warranty
These five models allow you to dive deeply into muscular anatomy. Begin with he Ward's muscle models. Each model shows a different type of muscle (smooth, cardiac, and skeletal) bundles. Each types shows general and detailed anatomical structures including capillaries, motor endplates and connective tissue. Move next to the MICROanatomy which details a muscle fiber magnified about 10,000 times. Individual myofibrils are shown in cross and longitudinal section for comparison. Finally let your students learn how the myofibrils contract with the moveable sarcomere model. Each Z-Line disk containing thin fibers, can be moved and fixed in position with the central thick fibers.
Customers Who Bought This Also Bought