To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Description:Oxaloacetate Assay Kit
- Size:100 assays
- Method:OD570 nm, or FL530/585 nm
- Sample:Plasma, serum, tissue and culture media.
- Processing time:20 minutes
- Brand name:EnzyChrom™
- Cat. no.:75878-344
- Supplier No.:EOAA-100
Spécifications
A propos de cet article
For quantitative determination of oxaloacetate and evaluation of drug effects on its metabolism
- Sensitive and accurate. Linear detection range: 7 to 400 μM oxaloacetate for colorimetric assays and 1 to 40 μM for fluorimetric assays.
- Method: OD570nm, or FL530/585nm
- Samples: Plasma, serum, tissue and culture media
Oxaloacetate (OAA) is an intermediate in the citric acid cycle and participates in gluconeogenesis. OAA is formed by the oxidation of malate, by deamidation of aspartate or by condensation of CO2 with pyruvate or phosphoenolpyruvate. BioAssay Systems’ Oxaloacetate Assay Kit provides a simple, direct and automation-ready procedure for measuring oxaloacetate concentration. OAA is converted into pyruvate which is then oxidized with the conversion of the dye into a colored and fluorescent form. The color intensity of the oxidized dye at 570 nm or fluorescence intensity at λex/em = 530/585 nm is directly proportional to the oxaloacetate concentration in the sample.