Biologix Serological Pipettes
Supplier: Avantor
Certificates
About this item
Color coded polystyrene serological pippetes come in sizes ranging from 1 to 50 ml.
- Highly transparent polystyrene
- Six volumes available
- Individually wrapped packaging
- Sterile
- Ascending and descending graduations
These color-coded serological pipettes have ascending and descending graduations.
Certifications: ISO Certified: GB/T 19001-2008/ISO 9001:2008.
Customers Who Bought This Also Bought