Milk Dilution Bottles
Fournisseur: Corning
Certificats
A propos de cet article
Made of Corning soda lime glass.
- Wide mouth bottle for easy measuring or filling
- Uses a GPI 40-400 threaded phenolic screw cap with rubber liner
- Autoclaving at 121 °C
Used for general storage of solutions and tissue culture work.
Certifications: Meets the requirements for milk dilution bottles stated in the 'Standard Methods for the Examination of Dairy Products', published by the American Public Health Association.
Spécifications
- Catalog No:
- 89208-252
- 89208-248
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey