Phlebotomy Tray
# de catalogue 470229-850
Fournisseur: United Product & Instrument
Spécifications
- Description:Phlebotomy Tray
- Cat. no.:470229-850
Spécifications
A propos de cet article
Have Everything At Hand.
- Constructed of tough LDPE
- Durable and easy to clean
- Includes Tube-Cube™
A simple and compact design for the busy phlebotomist. The Flexi-Phleb has large carrying capacity without bulk. The tray provides a handy pull out drawer that is great for storing needles, bandages and other items. The top level accepts removable Tube-CUBE™ racks that secure a variety of tubes, ice cups for temperature sensitive samples, sharps containers, or whatever your specific activities demand. Back at the lab, merely exchange your collected samples in Tube-CUBE’s at Specimen Processing with empty ones and you are ready to go again!
Ordering information: Includes 4 Tube Cubes; 2 - 9 place, 1-4 place and 1- 1 place; and 2 Squeegee Squeezer. Some items in the product image are not included in purchase.
Customers Who Bought This Also Bought