Sample Wrapper, Thomas Scientific
# de catalogue 48956-142
Fournisseur: Thomas Scientific
Small Business Enterprise
Spécifications
- Description:Sample wrapper, ogg, black
- Cat. no.:48956-142
- Supplier No.:6514F70
Spécifications
A propos de cet article
Sample Wrapper absorbs heat quickly for infrared ignition; die-cut from unsized paper
Sample area 32 x 30 mm, with 8 x 38 mm extending strip as fuse for ignition.
Customers Who Bought This Also Bought