Graduated Cylinder, Chemglass
Fournisseur: Chemglass
Certificats
A propos de cet article
These large capacity cylinders are made of polypropylene
- Has double scale and hex base for stability
- Steam autoclavable at 121 °C
- Safer and more convenient
The handle is generously sized for large hands but with a slender enough grip for smaller hands. Hand opening is 3 1/2” x 1 1/8”
The handle on the Holdfast™ graduated cylinder makes pouring safer and more convenient.
Customers Who Bought This Also Bought