Filtering Flask, Heavy Wall, Chemglass
Supplier: Chemglass
Certificates
About this item
These heavy wall Erlenmeyer shaped filtering flasks have a standard taper outer joint and a serrated hose connection for attachment to vacuum source.
Customers Who Bought This Also Bought