A propos de cet article
Adding a syringe filtration step prior to injection not only helps to ensure more consistent and reliable results; it also helps protect delicate instruments and prolongs column life.
- Precise results in sensitive analysis of ionic analytes
- Certified for low-level IC interference by ICP analysis
- Hydrophilic membrane provides excellent flow rates and low binding coefficients
- Low affinity for binding drugs, ideal for dissolution testing procedures
- Able to filter solutions at temperatures up to 100 °C
Available in sizes and membrane types to meet any application need, Thermo Scientific™ Titan3™ PES patented Syringe Filters provide cleaner sample extracts by removing interfering materials and fine particles.
PES syringe filters have a well defined pore structure, giving effective retention of particles without excessive pore blockage. Compatible with a wide range of solvents. It is the membrane of choice for ion chromatography applications.
Low extractable membranes and housings ensure that no filter-borne contaminants are added to your sample. Performance-tested under actual ICP conditions. Integral 1 mm borosilicate glass pre-filter (30 mm devices) ensures greater confidence with high solids samples. Reinforcing ring provides greater strength to the housing to prevent leakage and bursting (maximum pressure for 30 mm devices 15 bar / 210 psi). Housings are manufactured from solvent-resistant, low-extractable polypropylene resins specifically selected for wide compatibility with common HPLC sample matrices.
Color coding makes it easy to select the correct membrane and pore size. Packed in re-usable rigid transparent color-coded containers. Syringe filters can be sterilized by autoclave at 121 °C for 15 minutes. Enhanced Luer lock inlet prevents leakage; outlet fitting is a standard size male Luer-slip fitting for ease of filtrate collection.