ULTRA-WARE® Filtration Caps, Kimble Chase
# de catalogue KT953915-0047
Fournisseur: DWK Life Sciences (KIMBLE)
Spécifications
- Cap Size:GL 45
- Ø:47 mm
- Description:Filteration Cap, Glass Filled PTFE
- Cat. no.:KT953915-0047
- Supplier No.:953915-0047
Spécifications
A propos de cet article
These Filtration caps made of glass-filled PTFE with a vacuum adapter for 1/4” ID tubing.
- Upper screw clamp holds a solvent pickup adapter or funnel
- Coarse fritted glass filter support is removable
Delivery: Supplied with a PTFE or propylene o-ring.
Customers Who Bought This Also Bought