Gas Evolution Measurement Apparatus, Chemglass
Fournisseur: Chemglass
Certificats
A propos de cet article
Gas evolution measurement apparatus is for the determination of concentration or purity of some active metals and metal hydrides.
- Convenient and accurate method
Certain active metals and metal hydrides react rapidly and quantiatively with suitable hydrolytic solvents to produce hydrogen gas. The leveling bulb capacity varies with the capacity of the buret and has a hose connection.
Ordering information: Flexible tubing must be ordered seperately. Supplied with a buret, -10 to 260° thermometer, a 24/40 thermometer adapter and a leveling bulb with a 24/40 top outer and a lower detachable hose connection.
Customers Who Bought This Also Bought